Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Fisherbrand™ Polyurethane Tractor Sit-Stand Stool Chrome - Medium Bench Height with Stationary Glides in Polyurethane
SCI_ED
Catalog No. S41257 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Black
Blue
Gray
Adjustable (Height):
20 to 28 in.
With Backrest:
No
With Tilt:
No
Yes
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Adjustable (Height) With Backrest With Tilt
S41257 Blue 20 to 28 in. No No
15-000-759 Gray 20 to 28 in. No Yes
S41256 Black 20 to 28 in. No No
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S41257
Fisherbrand™ Polyurethane Tractor Sit-Stand Stools in Chrome are height adjustable for 36 in. work surfaces & features ergonomic adjustments & stationary glides. Self-skinning polyurethane is chemical, soil, & puncture resistant, easily cleaned, & features breathable holes for added comfort.
  • Adjustable from 20-28 inches from the top of the seat to the floor
  • Features ergonomic adjustments such as seat height
  • Locking seat tilt
  • Cast-aluminum base
  • Stationary, non-marring glides suitable for hard floors
  • Chemical, soil, and puncture resistant polyurethane
Specifications
Product Type Tractor Sit/Stand Stool
With Lumbar Support No
Load Bearing Capacity (English) 300 lb.
Adjustable (Height) 20 to 28 in.
Base Material Polished Cast Aluminum
With Armrest No
With Casters No
Footrest No
With Tilt No
Adjustable Yes
Color Blue
Load Bearing Capacity (Metric) 136 kg
Weight (English) 25 lb.
Foot Type Black-non-marring bell glides (suitable for both hard and carpeted floors)
With Backrest No
Material Polyurethane
Seating Type Stool
Show More Show Less
Product Title
Select an issue
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.